EMAIL THIS PAGE TO A FRIEND
266833 Aldrich

Iridium

foil, thickness 0.07 mm, 99.5% trace metals basis

Purchase

Properties

Related Categories Iridium, Materials Science, Metal and Ceramic Science, Metals
InChI Key   GKOZUEZYRPOHIO-UHFFFAOYSA-N
assay   99.5% trace metals basis
form   foil
resistivity   4.71 μΩ-cm
thickness   0.07 mm
bp   4130 °C(lit.)
mp   2450 °C(lit.)
density   22.65 g/cm3(lit.)

Description

Quantity

750 mg = 25 × 25 mm

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?