EMAIL THIS PAGE TO A FRIEND
269824 Aldrich

Methanol-d4

99.8 atom % D

Synonym: Methyl-d3 alcohol d, Tetradeuteromethanol

Purchase

Properties

Related Categories Methanol-d4, NMR Solvents, Solvent by Application, Solvents, Spectroscopy Solvents (IR, NMR, UV/Vis) More...
InChI Key   OKKJLVBELUTLKV-MZCSYVLQSA-N
isotopic purity   99.8 atom % D
assay   99% (CP)
packaging   pk of 10 × 1 mL
refractive index   n20/D 1.326(lit.)
bp   65.4 °C(lit.)
density   0.888 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
2
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C
Protocols & Articles

Related Content

Properties of Solvents Table

a. Gant, R., LC/GC 10(7), 515 (1992) 1. At 20 °C 2. At 20 °C +/- 5 °C relative to water at 4 °C 3. At 10mmHg pressure 4. At l=589.32nm
Keywords: Gas chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

accessory

Product #

Description

Add to Cart

Z693847 Scienceware® Break-Safe ampule opener

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?