EMAIL THIS PAGE TO A FRIEND
269883 Aldrich

Dichloromethane-d2

99.5 atom % D, contains 1 % (v/v) TMS

Synonym: Dideuteromethylenechloride, Methylene chloride-d2

Purchase

Properties

Related Categories Dichloromethane (Methylene chloride), Dichloromethane-d2, NMR Solvents, Solvent by Type, Solvent by Application,
vapor pressure   25.72 psi ( 55 °C)
  7.61 psi ( 20 °C)
InChI Key   YMWUJEATGCHHMB-DICFDUPASA-N
isotopic purity   99.5 atom % D
contains   1 % (v/v) TMS
refractive index   n20/D 1.422(lit.)
bp   40 °C(lit.)
density   1.362 g/mL at 25 °C(lit.)
mass shift   M+2

Price and Availability


Added X
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Blood, Central nervous system, Liver
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
2
Protocols & Articles

Related Content

Properties of Solvents Table

a. Gant, R., LC/GC 10(7), 515 (1992) 1. At 20 °C 2. At 20 °C +/- 5 °C relative to water at 4 °C 3. At 10mmHg pressure 4. At l=589.32nm
Keywords: Gas chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

accessory

Product #

Description

Add to Cart

Z693847 Scienceware® Break-Safe ampule opener

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?