• USA Home
  • 270024 - 1-Chloro-3-nitrobenzene

EMAIL THIS PAGE TO A FRIEND
270024 Aldrich

1-Chloro-3-nitrobenzene

95%

  • CAS Number 121-73-3

  • Linear Formula ClC6H4NO2

  • Molecular Weight 157.55

  •  Beilstein Registry Number 1907947

  •  MDL number MFCD00007202

  •  PubChem Substance ID 329752895

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitro Compounds, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   KMAQZIILEGKYQZ-UHFFFAOYSA-N
assay   95%
bp   236 °C(lit.)
mp   43-47 °C(lit.)
density   1.534 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?