• USA Home
  • 272914 - Trichloromethyl chloroformate

EMAIL THIS PAGE TO A FRIEND
272914 Aldrich

Trichloromethyl chloroformate

Synonym: Diphosgene, di-Phosgene

  • CAS Number 503-38-8

  • Linear Formula ClCOOCCl3

  • Molecular Weight 197.83

  •  Beilstein Registry Number 970225

  •  EC Number 207-965-9

  •  MDL number MFCD00015553

Purchase

Properties

InChI Key   HCUYBXPSSCRKRF-UHFFFAOYSA-N
bp   20 °C/10 mmHg(lit.)

Description

Application

Reactant for preparation of:
• Cyclic carbamimidates using a monophosphine gold(i) catalyst
• N-Alkenyl and cycloalkyl carbamates as dual acting histamine H3 and H4 receptor ligands
• Prostate-specific membrane antigen-targeted anticancer prodrugs
• Potential west nile virus protease inhibitors
• Antibody-drug conjugates (ADCs)
• Erythromycin A derivatives

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3277 8(6.1) / PGII
WGK Germany 
3
RTECS 
LQ7350000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?