• USA Home
  • 274267 - Amberlite® IRP-64 hydrogen form

EMAIL THIS PAGE TO A FRIEND
274267 Aldrich

Amberlite® IRP-64 hydrogen form

hydrogen form

Synonym: Polyacrilex Resin

Purchase

Properties

Related Categories Amberlite, Analytical/Chromatography, Cation, Cation Exchange Media, LPLC Separation Media,
description   Pharmaceutical Grade
moisture   ~10%
matrix   methacrylic-divinylbenzene (macroreticular)
matrix active group   weak acid functional group
particle size   100-400 mesh
capacity   ~10 meq/g by dry weight
compatibility   mode of use excipient

Description

Application

Weakly acidic cation exchange resin for taste masking, drug stabilization, carrier for cationic drugs, controlled release formulations (such as a carrier for certain basic (cationic) drugs and related substances), etc.
For Research Use Only.

Physical properties

Particle size:
>750 μm 15.0 to 30.0 %
>1500 μm 1.0 % maximum.
Sodium content: 0.20 % max
Heavy metals: 10 ppm max.
Residual methacrylic acid: 200 ppm max.
Loss on drying 5.0 % max.
Dry, white powder.

Legal Information

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Selection Guide for Polymeric Resins in Chromatography & Purification

Sigma-Aldrich carries a vast array of resins, from leading manufacturers; such as, Dow Chemical, Rohm & Haas, Mitsubishi, and Lanxess. Our product line includes adsorbent, ion-exchange (anion / catio...
Keywords: Adsorption, Antibiotics, Detergents, Fermentation, Filtration, Hormones, Ion Exchange, Pharmaceutical, Purification, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?