• USA Home
  • 274283 - Metanephrine hydrochloride

EMAIL THIS PAGE TO A FRIEND
274283 Aldrich

Metanephrine hydrochloride

99%

Synonym: 4-Hydroxy-3-methoxy-α-(methylaminomethyl)benzyl alcohol hydrochloride

Purchase

Properties

Related Categories Amino Alcohols, Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds More...
InChI Key   HRIQFVCFOPJYEQ-UHFFFAOYSA-N
assay   99%
mp   175 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DA4798000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?