• USA Home
  • 274453 - Ethyl 3-indoleacetate

EMAIL THIS PAGE TO A FRIEND
274453 Aldrich

Ethyl 3-indoleacetate

99%

Synonym: 3-[(Ethoxycarbonyl)methyl]indole, AA ethyl ester, Ethyl β-indolylacetate, Ethyl 2-(indol-3-yl)acetate, Ethyl indol-3-ylacetate, NSC 38002

Purchase

Properties

Related Categories Building Blocks, C12, Chemical Synthesis, Heterocyclic Building Blocks, Indoles More...
InChI Key   HUDBDWIQSIGUDI-UHFFFAOYSA-N
assay   99%
bp   164-166 °C/1 mmHg(lit.)
mp   44-45 °C(lit.)

Description

Packaging

5 g in glass bottle

Application

Reactant for preparation of:
• Pesticides
• Anti-inflammatory agents
• Antibacterial and antifungal agents
• Anticonvulsant agents
• Hypotensive agents
• Analgesic agents

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
NL3528000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles

Articles

MSMLS Online Plate Map

Metabolite descriptors included with the software download upon purchase of the product include: Name, Parent CID, molecular formula, molecular weight, CAS, ChEBI, HMDB ID, PubChem Compound and Subst...
Keywords: Mass spectrometry, Metabolites

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?