• USA Home
  • 274690 - Nafion® perfluorinated resin, powder

EMAIL THIS PAGE TO A FRIEND
274690 Aldrich

Nafion® perfluorinated resin, powder

35-60 mesh

Purchase

Properties

eq. wt.   1,100
particle size   35-60 mesh

Description

Physical form

Hydrogen ion form.

Legal Information

Nafion is a registered trademark of The Chemours Company FC, LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?