• USA Home
  • 275360 - 1-(2-Pyrimidyl)piperazine dihydrochloride

EMAIL THIS PAGE TO A FRIEND
275360 Aldrich

1-(2-Pyrimidyl)piperazine dihydrochloride

98%

Synonym: 2-(1-Piperazinyl)pyrimidine dihydrochloride, 2-Piperazinopyrimidine dihydrochloride

Purchase

Properties

Related Categories Building Blocks, C4 to C8, Chemical Synthesis, Heterocyclic Building Blocks, Piperazines More...
InChI Key   ZNZGJSLHXOMREP-UHFFFAOYSA-N
assay   98%
mp   282-287 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

New Trifl uoroborate Salts for Suzuki Coupling from Aldrich R&D

Trifluoroborates are air-stable alternatives to boronic acids in palladium-catalyzed Suzuki–Miyaura cross-coupling reactions.1,2 They are more robust, easier to handle, and less prone to protodeboron...
Aldrich ChemFiles 2005, 5.5, 11.
Keywords: Catalysis, Chemfiles, Coupling reactions, Cross couplings

Related Products

comparable product

Product #

Description

Add to Cart

83125 1-(2-Pyrimidyl)piperazine dihydrochloride, ≥96.0% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?