• USA Home
  • 275484 - trans-2,3-Dimethoxycinnamic acid

EMAIL THIS PAGE TO A FRIEND
275484 Aldrich

trans-2,3-Dimethoxycinnamic acid

97%

Purchase

Properties

Related Categories Building Blocks, C11 to C12, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   QAXPUWGAGVERSJ-VOTSOKGWSA-N
assay   97%
mp   182-184 °C(lit.)

Description

Packaging

5 g in glass bottle

Application

trans-2,3-Dimethoxycinnamic acid was used in the synthesis of (2E,2′E,2′′E)-N,N′,N′′-(nitrilotri-2,1-ethanediyl)tris[3-(2,3-dimethoxyphenyl)-2-propenamide].

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?