EMAIL THIS PAGE TO A FRIEND
276901 Aldrich

Disodium cyanamide

95%

Synonym: Cyanamide disodium salt, di-Sodium cyanamide

  • CAS Number 20611-81-8

  • Linear Formula NCNNa2

  • Molecular Weight 86.00

  •  Beilstein Registry Number 3687732

Purchase

Properties

InChI Key   OZABOWUUHUPMRP-UHFFFAOYSA-N
assay   95%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?