• USA Home
  • 276979 - Sodium perchlorate hydrate

EMAIL THIS PAGE TO A FRIEND
276979 Aldrich

Sodium perchlorate hydrate

≥99% (Purity determined on metals basis)

  • Linear Formula NaClO4 · xH2O

  • Molecular Weight 122.44 (anhydrous basis)

  •  EC Number 231-511-9

  •  MDL number MFCD00149164

  •  PubChem Substance ID 329753016

Purchase

Properties

Related Categories Essential Chemicals, Materials Science, Metal and Ceramic Science, Research Essentials, Salts,
InChI Key   IXGNPUSUVRTQGW-UHFFFAOYSA-M
assay   ≥99% (Purity determined on metals basis)
composition   Degree of hydration, ≤1

Price and Availability


Ultra-High Purity Inorganics
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1502 5.1 / PGII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

381225 Sodium perchlorate hydrate, 99.99% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?