• USA Home
  • 277037 - (1R,2R)-(+)-1-Phenylpropylene oxide

EMAIL THIS PAGE TO A FRIEND
277037 Aldrich

(1R,2R)-(+)-1-Phenylpropylene oxide

97%

Synonym: (2R,3R)-(+)-2-Methyl-3-phenyloxirane

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Epoxides, Organic Building Blocks More...
InChI Key   YVCOJTATJWDGEU-APPZFPTMSA-N
assay   97%
optical activity   [α]20/D +110°, neat
optical purity   ee: 99% (GLC)
refractive index   n20/D 1.518(lit.)
bp   201-207 °C(lit.)
density   1.006 g/mL at 25 °C(lit.)

Description

Packaging

1 g in glass bottle

Application

Chiral building block. Tool for studying enzyme hydrolysis rates.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DA0177000
Flash Point(F) 
156.2 °F
Flash Point(C) 
69 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?