• USA Home
  • 27832 - Poly(styrene-co-divinylbenzene)

EMAIL THIS PAGE TO A FRIEND
27832 Aldrich

Poly(styrene-co-divinylbenzene)

98% styrene – 2% DVB

Synonym: Polystyrene crosslinked with divinylbenzene

Purchase

Properties

Related Categories Chemical Synthesis, Other Resins, Solid Supported Synthesis, Specialty Synthesis
particle size   100-200 mesh

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?