• USA Home
  • 278483 - 1,2-Bis(4,4-dimethyl-2-oxazolin-2-yl)ethane

EMAIL THIS PAGE TO A FRIEND
278483 Aldrich

1,2-Bis(4,4-dimethyl-2-oxazolin-2-yl)ethane

97%

Synonym: 2,2′-(1,2-Ethanediyl)bis[4,5-dihydro-4,4-dimethyl]oxazole

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Other Ligands More...
InChI Key   UFUAASKHUNQXBP-UHFFFAOYSA-N
assay   97%
mp   55-58 °C(lit.)

Description

Packaging

1, 5 g in glass bottle

Application

Reacts with copper dichloride

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?