• USA Home
  • 279854 - Tricarbonyl(mesitylene)chromium(0)

EMAIL THIS PAGE TO A FRIEND
279854 Aldrich

Tricarbonyl(mesitylene)chromium(0)

98%

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Chromium More...
InChI Key   NIONDGZOEVKXEP-UHFFFAOYSA-N
assay   98%
mp   170-172 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?