• USA Home
  • 280860 - 11-Cyanoundecanoic acid

EMAIL THIS PAGE TO A FRIEND
280860 Aldrich

11-Cyanoundecanoic acid

97%

  • CAS Number 5810-18-4

  • Empirical Formula (Hill Notation) C12H21NO2

  • Molecular Weight 211.30

  •  PubChem Substance ID 329753037

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   YKHZGLKQBYTRHO-UHFFFAOYSA-N
assay   97%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?