EMAIL THIS PAGE TO A FRIEND
28206 Aldrich

Z-D-β-Dab(Boc)-OH

≥98.0% (TLC)

Synonym: (S)-4-(Boc-amino)-3-(Z-amino)butyric acid, Nβ-Z-Nγ-Boc-D-3,4-diamino-buttersäure, Nγ-Boc-Nβ-Z-D-3,4-diaminobutyric acid, Z-D-β-Dbu(Boc)-OH

  • CAS Number 96186-30-0

  • Empirical Formula (Hill Notation) C17H24N2O6

  • Molecular Weight 352.38

  •  MDL number MFCD03788644

  •  PubChem Substance ID 57648292

Purchase

Properties

Related Categories Beta-Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   KUMMZCONPFBQTC-ZDUSSCGKSA-N
assay   ≥98.0% (TLC)

Description

Other Notes

Building block for emeriamine

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

β-Amino Acids and Homologs

Although they are less abundant than their α-analogues, β-amino acids occur in nature both in free form and bound to peptides. Oligomers composed exclusively of β-amino acids (so-called β-peptides) m...
Matthias Junkers
Aldrich ChemFiles 2008, 8.7, 11.
Keywords: Chemfiles, Degradations, Metabolism, Peptidomimetics

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?