• USA Home
  • 283002 - Methyl-13C-triphenylphosphonium iodide

EMAIL THIS PAGE TO A FRIEND
283002 Aldrich

Methyl-13C-triphenylphosphonium iodide

99 atom % 13C

Synonym: 13C Labeled Methyltriphenylphosphonium iodide

Purchase

Properties

Related Categories Alphabetical Listings, C-C Bond Formation, Chemical Synthesis, M, Olefination,
InChI Key   JNMIXMFEVJHFNY-YTBWXGASSA-M
isotopic purity   99 atom % 13C
mp   186-188 °C(lit.)
mass shift   M+1

Description

Packaging

1 g in glass bottle

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Price and Availability


Stable Isotope
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?