EMAIL THIS PAGE TO A FRIEND
285269 Aldrich

(+)-Bicuculline

98%

  • CAS Number 485-49-4

  • Empirical Formula (Hill Notation) C20H17NO6

  • Molecular Weight 367.35

  •  Beilstein Registry Number 98786

  •  MDL number MFCD00005006

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   IYGYMKDQCDOMRE-ZWKOTPCHSA-N
assay   98%

Description

Biochem/physiol Actions

(+)-Bicuculline is a GABAA receptor antagonist.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?