• USA Home
  • 285374 - Chlorpromazine hydrochloride

EMAIL THIS PAGE TO A FRIEND
285374 Aldrich

Chlorpromazine hydrochloride

98%

Synonym: 2-Chloro-10-(3-dimethylaminopropyl)phenothiazine hydrochloride, CPZ, Largactil

  • CAS Number 69-09-0

  • Empirical Formula (Hill Notation) C17H19ClN2S · HCl

  • Molecular Weight 355.33

  •  Beilstein Registry Number 3779989

  •  MDL number MFCD00012654

  •  PubChem Substance ID 329753232

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   FBSMERQALIEGJT-UHFFFAOYSA-N
assay   98%

Description

Application

Substitute for benzidine, o-dianisidine, and o-tolidine in the determination of microquantities of hemoglobin and peroxidase.

Biochem/physiol Actions

Chlorpromazine demonstrates cytotoxic and antiproliferative activity against leukemic cells, but does not affect the viability of normal lymphocytes.

Phenothiazine antipsychotic; D2 dopamine receptor antagonist, H1 histamine receptor antagonist; inhibits calmodulin-dependent stimulation of cyclic nucleotide phosphodiesterase and nitric oxide synthase.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?