• USA Home
  • 285390 - 5β-Cholestan-3α-ol

EMAIL THIS PAGE TO A FRIEND
285390 Aldrich

5β-Cholestan-3α-ol

≥95%

Synonym: Epicoprostanol

Purchase

Properties

InChI Key   QYIXCDOBOSTCEI-KKFSNPNRSA-N
assay   ≥95%

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?