Synonym: 3α-Hydroxy-5β-androstan-17-one, 5β-Androsterone, Etiocholanone
CAS Number 53-42-9
Empirical Formula (Hill Notation) C19H30O2
Molecular Weight 290.44
Beilstein Registry Number 2217625
MDL number MFCD00064133
Popular Documents: FTNMR (PDF)
| assay | 98% |
| drug control | regulated under CDSA - not available from Sigma-Aldrich Canada |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?