• USA Home
  • 286109 - 19-Hydroxy-4-androsten-3,17-dione

EMAIL THIS PAGE TO A FRIEND
286109 Aldrich

19-Hydroxy-4-androsten-3,17-dione

Synonym: 19-Hydroxy-4-androstene-3,17-dione, 4-Androsten-19-ol-3,17-dione, 4-Androstene-3,17-dione-19-ol

Purchase

Properties

InChI Key   XGUHPTGEXRHMQQ-BGJMDTOESA-N
mp   168-169 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?