EMAIL THIS PAGE TO A FRIEND
286575 Aldrich

2-Methoxyestradiol

98%

Synonym: 1,3,5(10)-Estratriene-2,3,17-triol 2-methyl ether, 2,3,17β-Trihydroxy-1,3,5(10)-estratriene 2-methyl ether, 2-Methoxyestra-1,3,5(10)-triene-3,17β-diol, 2-Hydroxyestradiol 2-methyl ether, 3,17β-Dihydroxy-2-methoxy-1,3,5(10)-estratriene

  • CAS Number 362-07-2

  • Empirical Formula (Hill Notation) C19H26O3

  • Molecular Weight 302.41

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Alcohols, Building Blocks, C11 to C30+, C16 to C21, Chemical Synthesis,
InChI Key   CQOQDQWUFQDJMK-VKYAZVNUSA-N
assay   98%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN2811 - class 6.1 - PG 3 - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
KG7537500

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?