• USA Home
  • 28785 - 1,2-Epoxycyclododecane

EMAIL THIS PAGE TO A FRIEND
28785 Aldrich

1,2-Epoxycyclododecane

(cis+trans), ≥95% (GC)

Synonym: Cyclododecene oxide

  • CAS Number 286-99-7

  • Empirical Formula (Hill Notation) C12H22O

  • Molecular Weight 182.30

  •  Beilstein Registry Number 106689

  •  EC Number 206-012-4

  •  MDL number MFCD00005116

  •  PubChem Substance ID 24857298

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Epoxides, Organic Building Blocks, Oxygen Compounds More...
InChI Key   VLJLXEKIAALSJE-UHFFFAOYSA-N
assay   ≥95% (GC)
refractive index   n20/D 1.481
bp   274-276 °C(lit.)
density   0.947 g/mL at 20 °C(lit.)

Description

Packaging

100 mL in glass bottle

General description

The kinetics of the rearrangement of 1,2-epoxycyclododecane, catalyzed by lithium bromide in the presence of polyethylene glycol 400, was studied.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?