• USA Home
  • 288969 - tert-Butyl 4-acetyl-3,5-dimethyl-2-pyrrolecarboxylate

EMAIL THIS PAGE TO A FRIEND
288969 Aldrich

tert-Butyl 4-acetyl-3,5-dimethyl-2-pyrrolecarboxylate

98%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Pyrroles
InChI Key   ANUXYJVRWSTYSY-UHFFFAOYSA-N
assay   98%
mp   151-153 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?