• USA Home
  • 29095 - Cyclohexano-15-crown-5

EMAIL THIS PAGE TO A FRIEND
29095 Aldrich

Cyclohexano-15-crown-5

≥97.0%

  • CAS Number 17454-48-7

  • Empirical Formula (Hill Notation) C14H26O5

  • Molecular Weight 274.35

  •  Beilstein Registry Number 1682984

  •  EC Number 241-471-4

  •  MDL number MFCD00054545

Purchase

Properties

Related Categories Chelation/Complexation Compounds, Chemical Synthesis, Crown Ethers, Synthetic Reagents
InChI Key   GQRWGIWRQMNZNT-UHFFFAOYSA-N
assay   ≥97.0%
density   1.12 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
233.6 °F
Flash Point(C) 
112 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?