• USA Home
  • 293113 - Sulfur monochloride solution

EMAIL THIS PAGE TO A FRIEND
293113 Aldrich

Sulfur monochloride solution

1.0 M in methylene chloride

  • CAS Number 10025-67-9

  • Empirical Formula (Hill Notation) Cl2S2

  • Molecular Weight 135.04

  •  MDL number MFCD00011446

Purchase

Properties

Related Categories Essential Chemicals, Materials Science, Metal and Ceramic Science, Research Essentials, Salts,
vapor density   4.7 (vs air)
vapor pressure   25.05 psi ( 55 °C)
  7.31 psi ( 20 °C)
InChI Key   PXJJSXABGXMUSU-UHFFFAOYSA-N
autoignition temp.   451 °F
concentration   1.0 M in methylene chloride

Description

Packaging

Packaged under nitrogen in Sure/Seal bottles

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Price and Availability


Ultra-High Purity Inorganics
Safety & Documentation

Safety Information

Signal word 
Danger
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN2922 - class 8 - PG 1 - EHS- -acidic - Corrosive liquids, toxic, n.o.s., HI: all
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?