EMAIL THIS PAGE TO A FRIEND
293288 Aldrich

1-Propanol

HPLC grade, ≥99.5%

Synonym: Propyl alcohol

Purchase

Properties

grade   HPLC grade
vapor density   2.1 (vs air)
vapor pressure   10 mmHg ( 147 °C)
  14.9 mmHg ( 20 °C)
InChI Key   BDERNNFJNOPAEC-UHFFFAOYSA-N
assay   ≥99.5%
autoignition temp.   700 °F
purified by   glass distillation
expl. lim.   13.7 %
impurities   <0.080% water
evapn. residue   <0.0005%
refractive index   n20/D 1.384(lit.)
bp   97 °C(lit.)
  97 °C
mp   −127 °C(lit.)
density   0.804 g/mL at 25 °C(lit.)
λ   H2O reference
UV absorption   λ: 210 nm Amax: 1.00
  λ: 225 nm Amax: 0.50
  λ: 250 nm Amax: 0.05
  λ: 300-400 nm Amax: 0.01

Description

Packaging

Packaged in glass bottles

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1274 - class 3 - PG 2 - n-Propanol
WGK Germany 
1
RTECS 
UH8225000
Flash Point(F) 
71.6 °F
Flash Point(C) 
22 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?