• USA Home
  • 296740 - 2-(2-Nitroethoxy)tetrahydropyran

EMAIL THIS PAGE TO A FRIEND
296740 Aldrich

2-(2-Nitroethoxy)tetrahydropyran

Synonym: Tetrahydro-2-(2-nitroethoxy)-2H-pyran

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Pyrans
InChI Key   KQDXRRAVDLIMGM-UHFFFAOYSA-N
refractive index   n20/D 1.459(lit.)
solubility   dichloromethane: soluble25 mg/mL, clear, yellow to orange
density   1.143 g/mL at 25 °C(lit.)

Description

Packaging

5 g in glass bottle

Application

2-(2-Nitroethoxy)tetrahydropyran may be used in chemical synthesis.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
118.4 °F
Flash Point(C) 
48 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?