Synonym: Methyl-12C-d3 alcohol-d
CAS Number 811-98-3
Linear Formula 12CD3OD
Molecular Weight 36.06
EC Number 212-378-6
MDL number MFCD00044637
PubChem Substance ID 24857874
Popular Documents: FTNMR (PDF)
| Related Categories | Alphabetical Listings, M, NMR - Buffers and Reagents, NMR Solvents and Reagents, Stable Isotopes More... |
| description | 13C-depleted |
| isotopic purity | 99.95 atom % 12C |
| 99.8 atom % D | |
| refractive index |
n |
| bp | 65.4 °C(lit.) |
| density | 0.888 g/mL at 25 °C(lit.) |
| mass shift | M+4 |
1, 5 g in ampule
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?