EMAIL THIS PAGE TO A FRIEND
298395 Aldrich

Nile Red

Synonym: Nile Blue A Oxazone

Purchase

Properties

InChI Key   VOFUROIFQGPCGE-UHFFFAOYSA-N
mp   203-205 °C(lit.)
absorption   λmax 553 nm

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

N3013 Nile Red, Technical grade

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?