• USA Home
  • 299014 - 1,14-Tetradecanediol

EMAIL THIS PAGE TO A FRIEND
299014 Aldrich

1,14-Tetradecanediol

97%

Synonym: Tetradecamethylene glycol

Purchase

Properties

Related Categories Alcohols, Materials Science, Monomers, Polymer Science
vapor pressure   9 mmHg ( 200 °C)
InChI Key   XLKZJJVNBQCVIX-UHFFFAOYSA-N
assay   97%
mp   85-90 °C(lit.)

Description

Packaging

100 mg in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?