• USA Home
  • 299235 - Nitric-15N acid solution

EMAIL THIS PAGE TO A FRIEND
299235 Aldrich

Nitric-15N acid solution

40 wt. % in H2O, 98 atom % 15N

Purchase

Properties

Related Categories Alphabetical Listings, N-O, Stable Isotopes More...
InChI Key   GRYLNZFGIOXLOG-OUBTZVSYSA-N
isotopic purity   98 atom % 15N
concentration   40 wt. % in H2O
bp   120.5 °C
density   1.422 g/mL at 25 °C
mass shift   M+1

Description

Packaging

1 mL in ampule

Price and Availability


Stable Isotope
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2031BFORB 8 / PGII
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?