• USA Home
  • 300446 - (2S)-(+)-Glycidyl 4-nitrobenzoate

EMAIL THIS PAGE TO A FRIEND
300446 Aldrich

(2S)-(+)-Glycidyl 4-nitrobenzoate

98%

Synonym: (S)-(+)-Glycidyl 4-nitrobenzoate, (S)-(+)-Oxirane-2-methanol 4-nitrobenzoate

  • CAS Number 115459-65-9

  • Empirical Formula (Hill Notation) C10H9NO5

  • Molecular Weight 223.18

  •  Beilstein Registry Number 87687

  •  MDL number MFCD00064490

  •  PubChem Substance ID 24858095

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Epoxides, Organic Building Blocks More...
InChI Key   MUWIANZPEBMVHH-VIFPVBQESA-N
assay   98%
optical activity   [α]19/D +37°, c = 1.08 in chloroform
mp   60-62 °C(lit.)
storage temp.   2-8°C

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
RR0511020
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?