• USA Home
  • 300500 - (2S,3S)-(−)-3-(4-Nitrophenyl)glycidol

EMAIL THIS PAGE TO A FRIEND
300500 Aldrich

(2S,3S)-(−)-3-(4-Nitrophenyl)glycidol

98%

Synonym: trans-(−)-2,3-Epoxy-3-(p-nitrophenyl)-1-propanol

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Epoxides, Organic Building Blocks More...
InChI Key   TVWSYFXHQPGITR-IUCAKERBSA-N
assay   98%

Description

Packaging

250 mg in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
RR0511100
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?