• USA Home
  • 301302 - 1,7-Dioxaspiro[5.5]undecane

EMAIL THIS PAGE TO A FRIEND
301302 Aldrich

1,7-Dioxaspiro[5.5]undecane

98%

Synonym: Olive-fly ketal

Purchase

Properties

InChI Key   GBBVHDGKDQAEOT-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.464(lit.)
bp   193 °C/750 mmHg(lit.)
density   1.02 g/mL at 25 °C(lit.)

Description

Packaging

500 mg in ampule

General description

1,7-Dioxaspiro[5.5]undecane is also referred as olean. It is the major component of female sex attractant pheromone blend of the olive fruit fly Dacus oleae. Inclusion complexes of β-cyclodextrin with 1,7-dioxaspiro[5.5]undecane were studied by semiempirical calculations.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
147.2 °F
Flash Point(C) 
64 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

D7151 1,7-Dioxaspiro[5.5]undecane, ≥97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?