EMAIL THIS PAGE TO A FRIEND
302228 Aldrich

3,5-Difluoroanisole

97%

Purchase

Properties

Related Categories Aryl Fluorinated Building Blocks, Building Blocks, C2 to C7, C7, Chemical Synthesis,
InChI Key   OTGQPYSISUUHAF-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.466(lit.)
density   1.234 g/mL at 25 °C(lit.)

Description

Packaging

5 g in glass bottle

General description

The structure and vibrations of 3,5-difluoroanisole in first electronically excited state were studied by mass-analyzed resonant two-photon ionization technique as well as the quantum chemical calculations.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
111.2 °F
Flash Point(C) 
44 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?