• USA Home
  • 302406 - 1-Benzyl-3-pyrroline

EMAIL THIS PAGE TO A FRIEND
302406 Aldrich

1-Benzyl-3-pyrroline

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Pyrrolines
InChI Key   LRFHKHHUKGZIGE-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.542(lit.)
bp   60 °C/1 mmHg(lit.)
density   0.837 g/mL at 25 °C(lit.)

Description

Packaging

5 g in glass bottle

Application

1-Benzyl-3-pyrroline was used in synthesis of (S)-1-benzyl-3-hydroxypyrrolidine.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3267 8 / PGII
WGK Germany 
3
Flash Point(F) 
197.6 °F
Flash Point(C) 
92 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?