• USA Home
  • 307017 - N-Ethyldicyclohexylamine

EMAIL THIS PAGE TO A FRIEND
307017 Aldrich

N-Ethyldicyclohexylamine

96%

Synonym: N,N-Dicyclohexylethylamine

Purchase

Properties

Related Categories Amines, Building Blocks, C14, Chemical Synthesis, Nitrogen Compounds,
InChI Key   XRKQMIFKHDXFNQ-UHFFFAOYSA-N
assay   96%
refractive index   n20/D 1.485(lit.)
bp   114-115 °C/3 mmHg(lit.)
density   0.91 g/mL at 20 °C(lit.)

Description

Packaging

25 g in glass bottle

Application

N-Ethyldicyclohexylamine was used in the synthesis of F-amines along with other products by undergoing incomplete fluorination and fragmentation reactions.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2735PSN1 8 / PGII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?