• USA Home
  • 307335 - 4,7,13,18-Tetraoxa-1,10-diazabicyclo[8.5.5]eicosane

EMAIL THIS PAGE TO A FRIEND
307335 Aldrich

4,7,13,18-Tetraoxa-1,10-diazabicyclo[8.5.5]eicosane

90%

Synonym: Kryptofix® 211

Purchase

Properties

Related Categories Chelation/Complexation Compounds, Chemical Synthesis, Crown Ethers, Synthetic Reagents
InChI Key   LVNQVIZBPSRXAN-UHFFFAOYSA-N
assay   90%
refractive index   n20/D 1.5057(lit.)
bp   130 °C/0.002 mmHg(lit.)
density   1.097 g/mL at 25 °C(lit.)

Description

Legal Information

Kryptofix is a registered trademark of Merck KGaA, Darmstadt, Germany

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

accessory

Product #

Description

Add to Cart

Z693847 Scienceware® Break-Safe ampule opener

related product

Product #

Description

Add to Cart

94791 SnapIT Ampule Opener, Regular, for 1-2ml, 5-10ml, 10-15ml ampoules
43917 SnapIT Ampule Opener, large, for 5-10ml, 10-15ml, 20-25ml ampoules

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?