• USA Home
  • 307343 - Trimethylvinylammonium hydroxide solution

EMAIL THIS PAGE TO A FRIEND
307343 Aldrich

Trimethylvinylammonium hydroxide solution

15 wt. % in H2O

Synonym: Neurine

  • CAS Number 463-88-7

  • Empirical Formula (Hill Notation) C5H13NO

  • Molecular Weight 103.16

  •  PubChem Substance ID 329753570

Purchase

Properties

InChI Key   NIPLIJLVGZCKMP-UHFFFAOYSA-M
concentration   15 wt. % in H2O

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?