• USA Home
  • 310522 - (−)-2,3-Butanediyl di(p-toluenesulfonate)

EMAIL THIS PAGE TO A FRIEND
310522 Aldrich

(−)-2,3-Butanediyl di(p-toluenesulfonate)

99%

Synonym: (−)-2,3-Butanediol di-p-tosylate

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Protected Alcohols More...
InChI Key   MFRBMNNZDFDJOF-HZPDHXFCSA-N
assay   99%
optical activity   [α]20/D −38°, c = 2 in chloroform
mp   67-69 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?