• USA Home
  • 316466 - tert-Butyl methyl ether

EMAIL THIS PAGE TO A FRIEND
316466 Aldrich

tert-Butyl methyl ether

PRA grade, ≥99%

Synonym: MTBE, Methyl tert-butyl ether

Purchase

Properties

Related Categories Amber Glass Bottles, PRA, Solvent Bottles, Solvent Packaging Options, Solvents More...
grade   PRA grade
vapor density   3.1 (vs air)
vapor pressure   3.92 psi ( 20 °C)
InChI Key   BZLVMXJERCGZMT-UHFFFAOYSA-N
assay   ≥99%
autoignition temp.   705 °F
expl. lim.   15.1 %
evapn. residue   <0.0001%
halogenated residue (GC/ECD)   <10 ng/L (as heptachlor epoxide)
refractive index   n20/D 1.369(lit.)
bp   55-56 °C(lit.)
  55-56 °C
density   0.74 g/mL at 25 °C(lit.)

Description

Application

Suitable for pesticide residue analysis

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2398 3 / PGII
WGK Germany 
1
RTECS 
KN5250000
Flash Point(F) 
-27.4 °F
Flash Point(C) 
-33 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?