• USA Home
  • 317586 - 1-(p-Tosyl)-3,4,4-trimethylimidazolidine

EMAIL THIS PAGE TO A FRIEND
317586 Aldrich

1-(p-Tosyl)-3,4,4-trimethylimidazolidine

98%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Imidazolines/Imidazolidines
InChI Key   PAPAFDSNHZAZHR-UHFFFAOYSA-N
assay   98%
mp   66-67 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?