EMAIL THIS PAGE TO A FRIEND
318574 Aldrich

Propane

tank for propane torch

  • CAS Number 74-98-6

  • Linear Formula CH3CH2CH3

  • Molecular Weight 44.10

  •  Beilstein Registry Number 1730718

  •  EC Number 200-827-9

  •  MDL number MFCD00009359

  •  PubChem Substance ID 24859134

Purchase

Properties

Related Categories Burners, Heating, Labware More...
vapor density   1.5 (vs air)
vapor pressure   190 psi ( 37.7 °C)
  8.42 atm ( 21.1 °C)
InChI Key   ATUOYWHBWRKTHZ-UHFFFAOYSA-N
autoignition temp.   842 °F
expl. lim.   9.5 %
bp   -42.1 °C(lit.)
mp   -188 °C(lit.)
density   0.564 g/mL at 20 °C(lit.)

Description

General description

Contains fuel for 15 hours

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1978 2.1
WGK Germany 
nwg
RTECS 
TX2275000
Flash Point(F) 
-155.2 °F
Flash Point(C) 
-104 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?