EMAIL THIS PAGE TO A FRIEND
320005 Aldrich

Glycerol

ACS reagent, ≥99.5%

Synonym: 1,2,3-Propanetriol, Glycerin

Purchase

Properties

grade   ACS reagent
vapor density   3.1 (vs air)
vapor pressure   <1 mmHg ( 20 °C)
InChI Key   PEDCQBHIVMGVHV-UHFFFAOYSA-N
assay   ≥99.5%
autoignition temp.   698 °F
impurities   H2SO4, passes test (darkened)
  subst. darkened by H2SO4, passes test
  ≤0.003% Chlorinated cpds.
  ≤0.05% fatty acid esters (butyric acid)
  ≤0.5% water
ign. residue   ≤0.005%
color   APHA: ≤10
refractive index   n20/D 1.474(lit.)
bp   182 °C/20 mmHg(lit.)
  182 °C/20 mmHg
mp   20 °C(lit.)
density   1.25 g/mL(lit.)
anion traces   sulfate (SO42-): ≤0.001%
cation traces   heavy metals: ≤2 ppm

Description

Application

Glycerol is used both in sample preparation and gel formation for polyacrylamide gel electrophoresis. Glycerol (5-10%) increases the density of a sample so that the sample will layer at the bottom of a gel’s sample well. Glycerol is also used to aid in casting gradient gels and as a protein stabilizer and storage buffer component.

Other Notes

Contains acrolein, glucose

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
MA8050000
Flash Point(F) 
320 °F
Flash Point(C) 
160 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?