• USA Home
  • 323365 - Pentaamminechlorocobalt(III) chloride

EMAIL THIS PAGE TO A FRIEND
323365 Aldrich

Pentaamminechlorocobalt(III) chloride

≥99.99% trace metals basis

Synonym: Cobalt(III) chloropentammine chloride

Purchase

Properties

Related Categories Cobalt Salts, Essential Chemicals, Materials Science, Metal and Ceramic Science, Research Essentials,
InChI Key   NSYALVBBDKTCLE-UHFFFAOYSA-K
assay   ≥99.99% trace metals basis
form   powder

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

298301 Pentaamminechlorocobalt(III) chloride, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?